Assay Details
| Gene Symbol: IFNL3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128496.1 | NP_001121968.1 | 2 - 3 | 241 | 58 | |
| GenBank mRNA | AM773754.1 | - | 2 - 3 | 241 | 58 | |
| EF587763.1 | - | 2 - 3 | 241 | 58 | ||
| EU399904.1 | - | 2 - 3 | 241 | 58 | ||
| Gene Symbol: IFNL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289181.2 | XP_015144667.1 | 2 - 3 | 1186 | 58 | |
| Gene Symbol: LOC112532795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025152519.1 | XP_025008287.1 | 1 - 2 | 276 | 58 | |
More Information
Set Membership: |
|

IFNL3A
Genomic Map