Assay Details
| Gene Symbol: PLPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130488.2 | NP_001123960.2 | 3 - 4 | 584 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLPP3| Gene Symbol: PLPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130488.2 | NP_001123960.2 | 3 - 4 | 584 | 111 | |
Set Membership: |
|