Assay Details
| Gene Symbol: RS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134975.2 | NP_001128447.2 | 4 - 5 | 346 | 84 | |
| XM_015302585.2 | XP_015158071.2 | 2 - 3 | 1280 | 84 | ||
| GenBank mRNA | EU924185.1 | - | 4 - 5 | 87 | ||
More Information
Set Membership: |
|

RS1
Genomic Map