Assay Details
| Gene Symbol: GLP1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135551.1 | NP_001129023.1 | 3 - 4 | 348 | 66 | |
| GenBank mRNA | EU770586.1 | - | 3 - 4 | 348 | 66 | |
| JN859076.1 | - | 3 - 4 | 301 | 66 | ||
| JN859077.1 | - | 3 - 4 | 301 | 66 | ||
| JN859078.1 | - | 3 - 4 | 301 | 66 | ||
| JN859079.1 | - | 3 - 4 | 301 | 66 | ||
| JN859080.1 | - | 3 - 4 | 301 | 66 | ||
| JN859081.1 | - | 3 - 4 | 301 | 66 | ||
| JN859082.1 | - | 3 - 4 | 301 | 66 | ||
| JN859083.1 | - | 3 - 4 | 294 | 66 | ||
More Information
Set Membership: |
|

GLP1R
Genomic Map