Assay Details
| Gene Symbol: GSPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135677.1 | NP_001129149.1 | 12 - 13 | 1683 | 76 | |
| XM_015294247.2 | XP_015149733.1 | 12 - 13 | 1385 | 76 | ||
More Information
Set Membership: |
|

GSPT2
Genomic Map