Assay Details
| Gene Symbol: DEAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137649.1 | NP_001131121.1 | 9 - 10 | 1738 | 55 | |
| XM_015286216.2 | XP_015141702.1 | 9 - 10 | 1738 | 55 | ||
| XM_015286217.2 | XP_015141703.1 | 9 - 10 | 1738 | 55 | ||
| XM_015286218.2 | XP_015141704.1 | 9 - 10 | 1738 | 55 | ||
| XM_025150659.1 | XP_025006427.1 | 9 - 10 | 1738 | 55 | ||
More Information
Set Membership: |
|

DEAF1
Genomic Map