Assay Details
| Gene Symbol: ART4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142255.1 | NP_001135727.1 | 1 - 2 | 75 | 92 | |
| GenBank mRNA | EF195232.1 | - | 1 - 2 | 104 | 92 | |
| EF626644.1 | - | 1 - 2 | 261 | 92 | ||
| EU048538.1 | - | 1 - 2 | 75 | 92 | ||
More Information
Set Membership: |
|

ART4
Genomic Map