Assay Details
Gene Symbol: DRD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142849.2 | NP_001136321.1 | 3 - 4 | 993 | 90 | |
GenBank mRNA | MF967039.1 | - | 3 - 4 | 1029 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
