Assay Details
| Gene Symbol: GJD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204582.1 | NP_989913.1 | 1 - 2 | 139 | 87 | |
| GenBank mRNA | AF458098.1 | - | 1 - 2 | 139 | 87 | |
| AY700221.1 | - | 1 - 2 | 66 | 87 | ||
More Information
Set Membership: |
|

GJD2
Genomic Map