Assay Details
| Gene Symbol: PPP2R2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030886.1 | NP_001026057.1 | 7 - 8 | 976 | 78 | |
| XM_015297313.2 | XP_015152799.1 | 9 - 10 | 1053 | 78 | ||
| XM_015297314.2 | XP_015152800.2 | 7 - 8 | 828 | 78 | ||
| XM_015297315.2 | XP_015152801.2 | 7 - 8 | 863 | 78 | ||
| GenBank mRNA | AJ719690.1 | - | 7 - 8 | 976 | 78 | |
More Information
Set Membership: |
|

PPP2R2A
Genomic Map