Assay Details
| Gene Symbol: PAPD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012950.1 | NP_001012968.1 | 12 | 3145 | 74 | |
| XM_015282279.2 | XP_015137765.2 | 12 | 4192 | 74 | ||
| XM_015282280.2 | XP_015137766.2 | 12 | 4189 | 74 | ||
| XM_015282281.2 | XP_015137767.2 | 13 | 4301 | 74 | ||
| XM_015282284.2 | XP_015137770.2 | 11 | 4063 | 74 | ||
| XM_015282285.2 | XP_015137771.2 | 11 | 4033 | 74 | ||
| XM_015282286.2 | XP_015137772.2 | 11 | 4030 | 74 | ||
| GenBank mRNA | AJ720115.1 | - | 12 | 3145 | 74 | |
| CR387160.1 | - | 1 | 990 | 74 | ||
More Information
Set Membership: |
|

PAPD7
Genomic Map