Assay Details
| Gene Symbol: KCNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030378.1 | NP_001025549.1 | 1650 | 102 | ||
| XM_025143680.1 | XP_024999448.1 | 1785 | 102 | |||
More Information
Set Membership: |
|

KCNA3
Genomic Map