Assay Details
| Gene Symbol: RASL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030702.1 | NP_001025873.1 | 1 | 444 | 91 | |
| GenBank mRNA | AY729886.1 | - | 1 | 444 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RASL10A| Gene Symbol: RASL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030702.1 | NP_001025873.1 | 1 | 444 | 91 | |
| GenBank mRNA | AY729886.1 | - | 1 | 444 | 91 | |
Set Membership: |
|