Assay Details
| Gene Symbol: TMEM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031466.2 | NP_001026637.1 | 2 | 849 | 97 | |
| GenBank mRNA | AJ719800.1 | - | 2 | 849 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM11| Gene Symbol: TMEM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031466.2 | NP_001026637.1 | 2 | 849 | 97 | |
| GenBank mRNA | AJ719800.1 | - | 2 | 849 | 97 | |
Set Membership: |
|