Assay Details
| Gene Symbol: SOX17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039326.1 | NP_001034415.1 | 2 | 1145 | 99 | |
| GenBank mRNA | AB197131.1 | - | 2 | 1145 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOX17| Gene Symbol: SOX17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039326.1 | NP_001034415.1 | 2 | 1145 | 99 | |
| GenBank mRNA | AB197131.1 | - | 2 | 1145 | 99 | |
Set Membership: |
|