Assay Details
| Gene Symbol: ISLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044667.1 | NP_001038132.1 | 2 | 1453 | 62 | |
| GenBank mRNA | AB194233.1 | - | 2 | 1453 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ISLR2| Gene Symbol: ISLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044667.1 | NP_001038132.1 | 2 | 1453 | 62 | |
| GenBank mRNA | AB194233.1 | - | 2 | 1453 | 62 | |
Set Membership: |
|