Assay Details
| Gene Symbol: PTGER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083365.1 | NP_001076834.1 | 2 | 1050 | 71 | |
| GenBank mRNA | EF200120.1 | - | 2 | 1050 | 71 | |
More Information
Set Membership: |
|

PTGER2
Genomic Map