Assay Details
| Gene Symbol: CNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142256.1 | NP_001135728.1 | 2 | 151 | 97 | |
| XM_025144242.1 | XP_025000010.1 | 2 | 239 | 97 | ||
| GenBank mRNA | AJ719988.1 | - | 2 | 151 | 97 | |
More Information
Set Membership: |
|

CNN2
Genomic Map