Assay Details
| Gene Symbol: HOXA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204139.1 | NP_989470.1 | 1 | 348 | 77 | |
| GenBank mRNA | AY030050.1 | - | 1 | 348 | 77 | |
| JF317546.1 | - | 1 | 344 | 77 | ||
More Information
Set Membership: |
|

HOXA13
Genomic Map