Assay Details
| Gene Symbol: S100A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204148.1 | NP_989479.1 | 2 | 251 | 80 | |
| GenBank mRNA | U76365.1 | - | 2 | 251 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

S100A6| Gene Symbol: S100A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204148.1 | NP_989479.1 | 2 | 251 | 80 | |
| GenBank mRNA | U76365.1 | - | 2 | 251 | 80 | |
Set Membership: |
|