Assay Details
| Gene Symbol: CLDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204202.1 | NP_989533.1 | 1 | 354 | 90 | |
| GenBank mRNA | AF334677.1 | - | 1 | 354 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_204202.1 transcripts.
Set Membership: |
|

CLDN3
Genomic Map