Assay Details
| Gene Symbol: FZD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204221.2 | NP_989552.1 | 1 | 2553 | 82 | |
| GenBank mRNA | AF224317.1 | - | 1 | 2553 | 82 | |
| AY283066.1 | - | 1 | 1715 | 82 | ||
More Information
Set Membership: |
|

FZD7
Genomic Map