Assay Details
| Gene Symbol: SOX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204309.1 | NP_989640.1 | 2 | 1354 | 99 | |
| GenBank mRNA | AF302936.1 | - | 2 | 1354 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOX18| Gene Symbol: SOX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204309.1 | NP_989640.1 | 2 | 1354 | 99 | |
| GenBank mRNA | AF302936.1 | - | 2 | 1354 | 99 | |
Set Membership: |
|