Assay Details
| Gene Symbol: ZBTB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204680.1 | NP_990011.1 | 2 | 651 | 92 | |
| GenBank mRNA | AF086831.1 | - | 2 | 651 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBTB7A| Gene Symbol: ZBTB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204680.1 | NP_990011.1 | 2 | 651 | 92 | |
| GenBank mRNA | AF086831.1 | - | 2 | 651 | 92 | |
Set Membership: |
|