Assay Details
| Gene Symbol: FOXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204770.1 | NP_990101.1 | 4 | 2053 | 122 | |
| GenBank mRNA | AF150749.1 | - | 4 | 2053 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXA2| Gene Symbol: FOXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204770.1 | NP_990101.1 | 4 | 2053 | 122 | |
| GenBank mRNA | AF150749.1 | - | 4 | 2053 | 122 | |
Set Membership: |
|