Assay Details
| Gene Symbol: HMGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205295.1 | NP_990626.2 | 4 | 451 | 88 | |
| GenBank mRNA | D14314.1 | - | 3 | 408 | 88 | |
| X63463.1 | - | 451 | 88 | |||
More Information
Set Membership: |
|

HMGB3
Genomic Map