Assay Details
| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205315.1 | NP_990646.1 | 4 | 2253 | 85 | |
| GenBank mRNA | X07011.1 | - | 2253 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBB| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205315.1 | NP_990646.1 | 4 | 2253 | 85 | |
| GenBank mRNA | X07011.1 | - | 2253 | 85 | ||
Set Membership: |
|