Assay Details
| Gene Symbol: PCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205470.1 | NP_990801.1 | 550 | 97 | ||
| GenBank mRNA | J05419.1 | - | 550 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCK2| Gene Symbol: PCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205470.1 | NP_990801.1 | 550 | 97 | ||
| GenBank mRNA | J05419.1 | - | 550 | 97 | ||
Set Membership: |
|