Assay Details
| Gene Symbol: HBBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205489.2 | NP_990820.1 | 3 | 572 | 100 | |
| GenBank mRNA | BU428052.1 | - | 3 | 572 | 100 | |
| DR426326.1 | - | 3 | 565 | 100 | ||
| J00860.1 | - | 3 | 555 | 100 | ||
More Information
Set Membership: |
|

HBBA
Genomic Map