Assay Details
| Gene Symbol: NMUR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318437.1 | NP_001305366.1 | 2 - 3 | 986 | 92 | |
| GenBank mRNA | HQ833217.1 | - | 2 - 3 | 986 | 92 | |
More Information
Set Membership: |
|

NMUR2
Genomic Map