Assay Details
| Gene Symbol: SYNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318453.1 | NP_001305382.1 | 1 - 2 | 158 | 100 | |
| GenBank mRNA | AJ719536.1 | - | 1 - 2 | 158 | 100 | |
More Information
Set Membership: |
|

SYNGR2
Genomic Map