Assay Details
Gene Symbol: TMEM8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318457.1 | NP_001305386.1 | 3 - 4 | 465 | 68 | |
GenBank mRNA | BX931401.1 | - | 3 - 4 | 442 | 68 | |
BX934605.2 | - | 3 - 4 | 451 | 68 | ||
KP230536.1 | - | 3 - 4 | 465 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
