Assay Details
| Gene Symbol: AHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319008.1 | NP_001305937.1 | 5 - 6 | 524 | 106 | |
| XM_015289246.2 | XP_015144732.1 | 4 - 5 | 486 | 106 | ||
| GenBank mRNA | KM289156.1 | - | 5 - 6 | 524 | 106 | |
More Information
Set Membership: |
|

AHR2
Genomic Map