Assay Details
| Gene Symbol: PMS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319022.1 | NP_001305951.1 | 7 - 8 | 839 | 75 | |
| XM_015294281.2 | XP_015149767.1 | 7 - 8 | 872 | 75 | ||
| GenBank mRNA | AJ720540.1 | - | 7 - 8 | 839 | 75 | |
More Information
Set Membership: |
|

PMS2
Genomic Map