Assay Details
| Gene Symbol: LHX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320906.1 | NP_001307835.1 | 3 - 4 | 674 | 53 | |
| XM_004934489.3 | XP_004934546.1 | 3 - 4 | 832 | 53 | ||
| GenBank mRNA | AB485718.1 | - | 3 - 4 | 674 | 53 | |
More Information
Set Membership: |
|

LHX5
Genomic Map