Assay Details
| Gene Symbol: EPB42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321559.1 | NP_001308488.1 | 7 - 8 | 1090 | 68 | |
| GenBank mRNA | CR353331.1 | - | 7 - 8 | 1094 | 68 | |
More Information
Set Membership: |
|

EPB42
Genomic Map