Assay Details
| Gene Symbol: ENTPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321564.1 | NP_001308493.1 | 6 - 7 | 786 | 97 | |
| XM_004939433.3 | XP_004939490.1 | 6 - 7 | 792 | 97 | ||
| XM_015281774.2 | XP_015137260.1 | 6 - 7 | 781 | 97 | ||
| XM_015281775.2 | XP_015137261.1 | 6 - 7 | 796 | 97 | ||
| XM_025147329.1 | XP_025003097.1 | 6 - 7 | 792 | 97 | ||
| XM_025147330.1 | XP_025003098.1 | 6 - 7 | 792 | 97 | ||
More Information
Set Membership: |
|

ENTPD3
Genomic Map