Assay Details
| Gene Symbol: GADD45G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044666.1 | NP_001038131.1 | 3 - 4 | 387 | 59 | |
| GenBank mRNA | BX934459.2 | - | 3 - 4 | 385 | 59 | |
| CR352538.1 | - | 3 - 4 | 474 | 59 | ||
| DQ358722.1 | - | 3 - 4 | 387 | 59 | ||
More Information
Set Membership: |
|

GADD45G
Genomic Map