Assay Details
| Gene Symbol: MAFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079489.1 | NP_001072957.1 | 1 - 2 | 67 | 88 | |
| XM_015295426.2 | XP_015150912.1 | 2 - 3 | 2593 | 88 | ||
| XM_015295427.2 | XP_015150913.1 | 2 - 3 | 200 | 88 | ||
| XM_015295428.2 | XP_015150914.1 | 2 - 3 | 210 | 88 | ||
| XM_015295429.2 | XP_015150915.1 | 2 - 3 | 360 | 88 | ||
| XM_015295430.2 | XP_015150916.1 | 2 - 3 | 571 | 88 | ||
| XM_015295431.2 | XP_015150917.1 | 2 - 3 | 1724 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001079489.1, XM_015295426.2, XM_015295427.2, XM_015295428.2, XM_015295429.2, XM_015295430.2, XM_015295431.2 transcripts.
Set Membership: |
|

MAFG
Genomic Map