Assay Details
Gene Symbol: EVI2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080893.1 | NP_001074362.1 | 1 - 2 | 128 | 67 | |
GenBank mRNA | AJ720220.1 | - | 1 - 2 | 128 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ720220.1, NM_001080893.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
