Assay Details
| Gene Symbol: GLP2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163248.1 | NP_001156720.1 | 10 - 11 | 1249 | 70 | |
| GenBank mRNA | FJ899744.1 | - | 10 - 11 | 1249 | 70 | |
More Information
Set Membership: |
|

GLP2R
Genomic Map