Assay Details
| Gene Symbol: SEPHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164084.3 | NP_001157556.1 | 7 - 8 | 875 | 100 | |
| XM_015296736.2 | XP_015152222.1 | 7 - 8 | 1375 | 100 | ||
| GenBank mRNA | CF256191.1 | - | 4 - 5 | 446 | 100 | |
| CR385500.1 | - | 6 - 7 | 637 | 100 | ||
| DR418560.1 | - | 6 - 7 | 834 | 100 | ||
More Information
Set Membership: |
|

SEPHS1
Genomic Map