Assay Details
| Gene Symbol: DYNC1LI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167737.1 | NP_001161209.1 | 12 - 13 | 1509 | 74 | |
| XM_015281493.2 | XP_015136979.1 | 11 - 12 | 1690 | 74 | ||
| GenBank mRNA | DR419552.1 | - | 8 - 9 | 847 | 74 | |
More Information
Set Membership: |
|

DYNC1LI1
Genomic Map