Assay Details
| Gene Symbol: HTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173440.1 | NP_001166911.1 | 2 - 3 | 892 | 78 | |
| XM_025142550.1 | XP_024998318.1 | 2 - 3 | 4439 | 78 | ||
| XM_025142551.1 | XP_024998319.1 | 2 - 3 | 4439 | 78 | ||
| XM_025142552.1 | XP_024998320.1 | 2 - 3 | 4439 | 78 | ||
| XM_025142553.1 | XP_024998321.1 | 2 - 3 | 4439 | 78 | ||
| GenBank mRNA | GU256248.1 | - | 2 - 3 | 892 | 78 | |
More Information
Set Membership: |
|

HTR6
Genomic Map