Assay Details
| Gene Symbol: CYP4F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329637.1 | NP_001316566.1 | 7 - 8 | 986 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP4F11| Gene Symbol: CYP4F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329637.1 | NP_001316566.1 | 7 - 8 | 986 | 82 | |
Set Membership: |
|