Assay Details
| Gene Symbol: SLC2A11L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347710.1 | NP_001334639.1 | 3 - 4 | 291 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A11L1| Gene Symbol: SLC2A11L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347710.1 | NP_001334639.1 | 3 - 4 | 291 | 71 | |
Set Membership: |
|