Assay Details
| Gene Symbol: CXCL13L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348655.1 | NP_001335584.1 | 3 - 4 | 312 | 118 | |
| GenBank mRNA | CR352598.1 | - | 1 - 2 | 1093 | 118 | |
More Information
Set Membership: |
|

CXCL13L3
Genomic Map