Assay Details
| Gene Symbol: SELENOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348769.1 | NP_001335698.1 | 4 - 5 | 619 | 55 | |
| GenBank mRNA | BX936186.2 | - | 4 - 5 | 610 | 55 | |
| KT206227.1 | - | 1 - 2 | 94 | 55 | ||
More Information
Set Membership: |
|

SELENOP2
Genomic Map