Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001353945.1 | NP_001340874.1 | 3 - 4 | 698 | 89 | |
| GenBank mRNA | S79838.1 | - | 2 - 3 | 506 | 89 | |
More Information
Set Membership: |
|

SERPINC1
Genomic Map