Assay Details
| Gene Symbol: RAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204104.2 | NP_989435.2 | 1 - 2 | 184 | 108 | |
| GenBank mRNA | AF420601.1 | - | 1 - 2 | 184 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_204104.2 transcripts.
Set Membership: |
|

RAX2
Genomic Map